Human adiponectin trimeric form (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store lyophilized Adiponectin at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized AdipoQ remains stable until the expiry date when stored at −20 °C.
- Biological activity:ED50= 3-8.5 ug/ml, as determined by its ability to inhibit proliferation of HASMCs induced by HB EGF.
- Protein synonyms:ACDC
- Protein/peptide name:Adiponectin, trimeric Form
- Purity:Greater than 95% as determined by SDS-PAGE.
- Solubility:Add deionized water to working volume of 0.5mg/ml and let the lyophilized pellet dissolve completely. Product is not sterile! Please filter the product by an appropriate sterile filter before using it in the cell culture.
- Sequence:ETTTQGPGVL LPLPKGAATG WMAGIPGHPG HNGAPGRDGR DGTPGEKGEK GDPGLIGPKG DIGETGVPGA EGPRGFPGIQ GRKGEPGEGA YVYRSAFSVG LETYVTIPNM PIRFTKIFYN QQNHYDGSTG KFHCNIPGLY YFAYHIVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTNDYKDD DDK.
- Formulation:Lyophilized from 0.5 mg/ml in 0.05 phosphate buffer, 0.05 NaCl, pH 7.2.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together