Order Entry
Italy
ContactUsLinkComponent
Human thyroid stimulating hormone (from pituitary glands)
Human thyroid stimulating hormone (from pituitary glands)
Catalog # BIRBORB80766-10
Supplier:  Biorbyt
CAS Number:  
Human thyroid stimulating hormone (from pituitary glands)
Catalog # BIRBORB80766-10
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Source:
    Pituitary glands
  • Species:
    Human
  • Storage conditions:
    TSH Human although stable at room temperature for weeks, should be stored at -15 °C.
  • Protein synonyms:
    Glycoprotein hormones alpha chain
  • Protein/peptide name:
    Thyroid Stimulating Hormone
  • Purity:
    Greater than 96.0% as determined by SDS-PAGE.
  • Solubility:
    It is recommended to reconstitute the lyophilized TSH in de-ionized water.
  • Formulation:
    Lyophilized from 0.05M NH4C O3 buffer, containing Trehalose at 25X hormone weight.
  • Tested applications:
    SDS-PAGE

Specifications

About this item