Human recombinant a-Internexin (from E. coli)
Catalog # BIRBORB80907-5
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Protein synonyms:Alpha-internexin
- Protein/peptide name:a-Internexin
- Formulation:-Internexin protein at 0.1mg/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS