Recombinant toxoplasma Gondii P30
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Storage conditions:Store at 4 °C. Upon arrival, store at −20 °C
- Protein synonyms:SAG1
- Protein/peptide name:Toxoplasma Gondii P30
- Purity:>95% pure as determined by 10% PAGE (coomassie staining).
- Formulation:50mM Tris pH 8, 1.5M urea and 50% glycerol
- Tested applications:ELISA , WB
Specifications
Frequently Bought Together