HIV Type-O Envelope (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Storage conditions:HIV type-O protein is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HIV Type-O Envelope
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
- Formulation:20 mM PBS pH-7.8, NaCl 0.5M, DTT 1mM, 8M urea 0.4M imidasole.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together