rHIV-1 p24 Biotin (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Storage conditions:HIV-1 p24 protein is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
- Protein/peptide name:rHIV-1 p24 Biotin
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
- Formulation:8M Urea, 20 mM Tris-HCl pH 8.0, 10mM beta mercaptoethanol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together