HCV NS3 genotype-1c
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HCV NS3 Genotype-1c
- Purity:HCV NS3 Genotype-1c protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV NS3 Genotype-1c protein was purified by proprietary chromatographic technique.
- Formulation:1.5M urea, 25mM Tris-HCl pH-8, 0.2% Triton-X 50% Glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together