Order Entry
Italy
ContactUsLinkComponent
Recombinant Borrelia p41
Recombinant Borrelia p41
Supplier:  Biorbyt
Certificates
Recombinant Borrelia p41
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein/peptide name:
    Borrelia p41
  • Purity:
    Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
  • Formulation:
    20 mM Tris-HCl, pH 7.5, 10mM beta mercaptoethanol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together