HIV-1 p17/ p24/ gp120/ gp41
Catalog # BIRBORB85298-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Source:
- Species:
- Environmentally Preferable:
- Storage conditions:Store at−20 °C.
- Protein synonyms:Immunodeficiency Virus 1 antigen
- Protein/peptide name:HIV-1 p17/ p24/ gp120/ gp41
- Purity:Greater than 95% by SDS-PAGE and HPLC
- Formulation:Liquid. Supplied in 20 mM PBS pH 7.8, 20 mM NaCl,1 mM DTT and 8 M urea.
- Tested applications:WB , SDS-PAGE