Order Entry
Italy
ContactUsLinkComponent
Human recombinant PPM1A (from E. coli)
Human recombinant PPM1A (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant PPM1A (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
  • Biological activity:
    8,000 U/mg.
  • Protein synonyms:
    Protein phosphatase 1A
  • Protein/peptide name:
    PPM1A
  • Purity:
    Greater than 95.0% as determined by(a)Analysis by RP-HPLC.(b)Analysis by SDS-PAGE.
  • Sequence:
    MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWILMGAF LDKPKMEKHN AQGQGNGLR YGLSSMQGWR VEMEDAHTAV IGLPSGLESW SFFAVYDGHA G SQVAKYCC EHLLDHITNN QDFKGSAGAP SVENVKNGIR TG FLEIDEH MRV MSEKKH GADRSGSTAV GVLISPQHTY FINCGDSRGL LCRNRKVHFF TQDH KPSNP LEKERIQNAG GSVMIQRVNG SLAVSRALGD FDYKCVHGKG PTEQL VSPE PEVHDI ERS EEDDQFI IL ACDGIWDVMG NEELCDFVRS SRLEVTDDL RLEVTDDL E KVCNEVVDTC LYKGSRDNMS VILICFPNAP KVSPEAVKKE AELDKYLEC RVEEIIKKQG EGVPDLVHV M RTLASENI PSLPPGGELA SKRNVIEAVY NR LNPYKND DTDSTSTDDM W.
  • Formulation:
    The protein (1mg/ml) In phosphate-buffered saline pH 7.4)
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together