Order Entry
Italy
ContactUsLinkComponent
Mouse EGF (from submaxillary glands)
Mouse EGF (from submaxillary glands)
Supplier:  Biorbyt
Certificates
Mouse EGF (from submaxillary glands)
Supplier:  Biorbyt

Specifications

  • Source:
    Submaxillary glands
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized Epidermal Growth Factor Mouse although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution EGF should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    The biological activity is measured in proliferation assay using BALB/MK cells.
  • Protein synonyms:
    Urogastrone
  • Protein/peptide name:
    EGF
  • Purity:
    Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Epidermal Growth Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution containing 0.01M sodium acetate buffer.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together