Order Entry
Italy
ContactUsLinkComponent
Recombinant SPA (from E. coli)
Recombinant SPA (from E. coli)
Supplier:  Biorbyt
Certificates
Recombinant SPA (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Storage conditions:
    SPA should be stored at −20 °C.
  • Biological activity:
    Greater than 95.0% binding activity to human IgG.
  • Protein synonyms:
    Immunoglobulin G-binding
  • Protein/peptide name:
    SPA
  • Purity:
    Greater than 98.0% as determined by RP-HPLC.
  • Formulation:
    The protein solution contains no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together