Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Rat calbindin 1 (from E. coli)

Marketsource
Nome del fornitore:   Biorbyt
Rat calbindin 1 (from <i>E. coli</i>)
Nome del fornitore:  Biorbyt
Marketsource
Certificates

2 Options Available Below

Product Details & Documents

 

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • BIRBORB80982-1
  • BIRBORB80982-5
  • Protein/peptide name:
  • Calbindin 1
  • Calbindin 1
  • Protein synonyms:
  • avian-type
  • avian-type
  • Species:
  • Rat
  • Rat
  • Source:
  • E. coli
  • E. coli
  • Purity:
  • Greater than 90% as determined by SDS-PAGE.
  • Greater than 90% as determined by SDS-PAGE.
  • Formulation:
  • The protein was lyophilized from concentrated solution (1mg/ml) containing 0.1mM CaCl2.
  • The protein was lyophilized from concentrated solution (1mg/ml) containing 0.1mM CaCl2.
  • Storage conditions:
  • Lyophilized CABP28K although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CABP28K should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Lyophilized CABP28K although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CABP28K should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Solubility:
  • It is recommended to reconstitute the lyophilized CABP28K in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • It is recommended to reconstitute the lyophilized CABP28K in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Tested applications:
  • SDS-PAGE
  • SDS-PAGE

Specifications

Product Family Options

Product Information

  • Pk
    Disponibilità
    Price
  • 1 µG
    Log in for availability
    Log in for availability
    1 * 1 µG
    1 * 1 µG

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BIRBORB80982-1
    Protein/peptide name
    Calbindin 1
    Protein synonyms
    avian-type
    Species
    Rat
    Source
    E. coli
    Purity
    Greater than 90% as determined by SDS-PAGE.
    Formulation
    The protein was lyophilized from concentrated solution (1mg/ml) containing 0.1mM CaCl2.
    Storage conditions
    Lyophilized CABP28K although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CABP28K should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
    Solubility
    It is recommended to reconstitute the lyophilized CABP28K in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
    Tested applications
    SDS-PAGE
    Marketsource
  • 5 µG
    Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BIRBORB80982-5
    Protein/peptide name
    Calbindin 1
    Protein synonyms
    avian-type
    Species
    Rat
    Source
    E. coli
    Purity
    Greater than 90% as determined by SDS-PAGE.
    Formulation
    The protein was lyophilized from concentrated solution (1mg/ml) containing 0.1mM CaCl2.
    Storage conditions
    Lyophilized CABP28K although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CABP28K should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
    Solubility
    It is recommended to reconstitute the lyophilized CABP28K in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
    Tested applications
    SDS-PAGE
    Marketsource