Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human FGF 19 (from
E. coli
)
Human FGF 19 (from E. coli)
Human FGF 19 (from E. coli)
Catalog # BIRBORB138859-50
Supplier:
Biorbyt
CAS Number:
Expand image
Human FGF 19 (from E. coli)
Catalog # BIRBORB138859-50
Supplier:
Biorbyt
CAS Number:
Specifications
Pk:
50 µG
Source:
E. coli
Species:
Human
Protein synonyms:
FGF19
UniProtKB:
O95750
Protein/peptide name:
FGF 19
Sequence:
Phe27-Lys216
Specifications
About this item
Product Details
Documentation
Product Details
Documentation