Rat IL4 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:B cell growth factor 1
- Protein/peptide name:IL4
- Purity:> 95 % by SDS-PAGE and HPLC analyses
- Molecular weight:14.0 kDa
- Sequence:HGCNDSPLRE IINTLNQVTE KGTPCTEMFV PDVLTATRNT TENELICRAS RVLRKFYFPRDVPPCLKNKS GVLGELRKLC RGVSGLNSLR SCTVNESTLT TLKDFLESLK SILRGKYLQSCTSMS
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 5% trehalose.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together