Order Entry
Italy
ContactUsLinkComponent
 
Human UBC5 (from E. coli)
 :  Biorbyt
 
Human UBC5 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Ubiquitin Conjugating Enzyme E2 D3
  • Protein/peptide name:
    UBC5
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    17.7 kDa
  • Sequence:
    MHHHHHHAMA LKRINKELSD LARDPPAQCS AGPVGDDMFH WQATIMGPNDSPYQGGVFFL TIHFPTDYPF KPPKVAFTTR IYHPNINSNG SICLDILRSQ WSPALTISKVLLSICSLLCD PNPDDPLVPE IARIYKTDRD KYNRISREWT QKYAM
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, 1mM DTT, pH 7.5.
  • Tested applications:
    SDS-PAGE , HPLC

 

 

Frequently Bought Together