Order Entry
Italy
ContactUsLinkComponent
TNF alpha (from Escherichia coli)
TNF alpha (from Escherichia coli)
Supplier:  Biorbyt
Certificates
TNF alpha (from Escherichia coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    APC1
  • Protein/peptide name:
    TNF alpha
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together