Order Entry
Italy
ContactUsLinkComponent
Rat IL13 (from E. coli)
Rat IL13 (from E. coli)
Supplier:  Biorbyt
Certificates
Rat IL13 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Allergic rhinitis
  • Protein/peptide name:
    IL13
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses
  • Molecular weight:
    12.1 kDa
  • Sequence:
    PVRRSTSPPV ALRELIEELS NITQDQKTSL CNSSMVWSVD LTAGGFCAAL ESLTNISSCNAIHRTQRILN GLCNQKASDV ASSPPDTKIE VAQFISKLLN YSKQLFRYGH
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together