Human HBEGF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Heparin-binding EGF-like growth factor
- Protein/peptide name:HBEGF
- Purity:> 97% by SDS-PAGE and HPLC analyses
- Molecular weight:9.7 kDa
- Sequence:DLQEADLDLL RVTLSSKPQA LATPNKEEHG KRKKKGKGLG KKRDPCLRKYKDFCIHGECK YVKELRAPSC ICHPGYHGER CHGLSL
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in 20mM PB, pH7.4, 130mM NaCl
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together