Mouse FGF8 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Murine Fibroblast Growth Factor-8
- Protein/peptide name:FGF8
- Purity:> 97% by SDS-PAGE and HPLC analyses
- Molecular weight:28.1kDa
- Sequence:QVRSAAQKRG PGAGNPADTL GQGHEDRPFG QRSRAGKNFT NPAPNYPEEGSKEQRDSVLP KVTQRHVREQ SLVTDQLSRR LIRTYQLYSR TSGKHVQVLA NKRINAMAEDGDPFAKLIVE TDTFGSRVRV RGAETGLYIC MNKKGKLIAK SNGKGKDCVF TEIVLENNYTALQNAKYEGW YMAFTRKGRP RKGSKTRQHQ REVHFMKRLP RGHHTTEQSLRFEFLNYPPF TRSLRGSQRT WAPEPR
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4 500mMNaCl.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together