Order Entry
Italy
ContactUsLinkComponent
Human sFasR (fromE. coli)
Human sFasR (fromE. coli)
Supplier:  Biorbyt
Certificates
Human sFasR (fromE. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein/peptide name:
    sFasR
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses
  • Molecular weight:
    17.6 kDa
  • Sequence:
    RLSSKSVNAQ VTDINSKGLE LRKTVTTVET QNLEGLHHDG QFCHKPCPPG ERKARDCTVNGDEPDCVPCQ EGKEYTDKAH FSSKCRRCRL CDEGHGLEVE INCTRTQNTK CRCKPNFFCNSTVCEHCDPC TKCEHGIIKE CTLTSNTKCK EEGSRSN
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together