Order Entry
Italy
ContactUsLinkComponent
Mouse DCIP-1 (from E. coli)
Mouse DCIP-1 (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse DCIP-1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Murine Dendritic Cell Inflammatory
  • Protein/peptide name:
    DCIP-1
  • Purity:
    > 97% by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    7.9 kDa
  • Sequence:
    AVVASELRCQ CLNTLPRVDF ETIQSLTVTP PGPHCTQTEV IATLKDGQEV CLNPQGPRLQIIIKKILKSG KSS
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together