Human CYR61 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Cysteine-rich Angiogenic
- Protein/peptide name:CYR61
- Purity:> 95 % by SDS-PAGE and HPLC analyses
- Molecular weight:39.4 kDa
- Sequence:TCPAACHCPL EAPKCAPGVG LVRDGCGCCK VCAKQLNEDC SKTQPCDHTKGLECNFGASS TALKGICRAQ SEGRPCEYNS RIYQNGESFQ PNCKHQCTCI DGAVGCIPLCPQELSLPNLG CPNPRLVKVT GQCCEEWVCD EDSIKDPMED QDGLLGKELG FDASEVELTRNNELIAVGKG SSLKRLPVFG MEPRILYNPL QGQKCIVQTT SWSQCSKTCG TGISTRVTNDNPECRLVKET RICEVRPCGQ PVYSSLKKGK KCSKTKKSPE PVRFTYAGCL SVKKYRPKYCGSCVDGRCCT PQLTRTVKMR FRCEDGETFS KNVMMIQSCK CNYNCPHANEAAFPFYRLFN DIHKFRD
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in citrate buffer solution, 300 mM NaCl, pH3.0.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together