Order Entry
Italy
ContactUsLinkComponent
TRP p41 mosaic GST
TRP p41 mosaic GST
Catalog # BIRBORB98098-100
Supplier:  Biorbyt
CAS Number:  
TRP p41 mosaic GST
Catalog # BIRBORB98098-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Storage conditions:
    Store at−20 °C.
  • Protein/peptide name:
    TRP p41 mosaicGST
  • Purity:
    Greater than 90% by SDS-PAGE
  • Formulation:
    Liquid. Supplied in 25mM Tris-HCl pH 8.0, 60mM NaCland 50% glycerol.
  • Tested applications:
    ELISA , WB

Specifications

About this item