Order Entry
Italy
ContactUsLinkComponent
 
HSV-II Ag (from E. coli)
  BIRBORB98960-1
 :  Biorbyt
 :  
HSV-II Ag (from E. coli)
  BIRBORB98960-1
 :  Biorbyt
 :  

 

  • Pk:
    1 mg
  • Source:
    E. coli
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    HSV-II Ag
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulation:
    PBS, pH 7.4
  • Tested applications:
    ELISA , WB , IHC-P , RIA