Order Entry
Italy
ContactUsLinkComponent
Recombinant cytokeratin 18
Recombinant cytokeratin 18
Catalog # BIRBORB98851-100
Supplier:  Biorbyt
CAS Number:  
Recombinant cytokeratin 18
Catalog # BIRBORB98851-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein synonyms:
    CYK 18
  • Protein/peptide name:
    Cytokeratin 18
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulation:
    PBS, pH 7.4
  • Tested applications:
    ELISA

Specifications

About this item