Order Entry
Italy
ContactUsLinkComponent
Rubella virus E1, Biorbyt
Rubella virus E1, Biorbyt
Catalog # BIRBORB79788-1
Supplier:  Biorbyt
CAS Number:  
Rubella virus E1, Biorbyt
Catalog # BIRBORB79788-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Storage conditions:
    Store at −20 °C.
  • Protein/peptide name:
    Rubella virus E1
  • Purity:
    Greater than 95% by SDS-PAGE gel analysis.
  • Molecular weight:
    8.6 kDa
  • Tested applications:
    ELISA , WB

Specifications

About this item