Order Entry
Italy
ContactUsLinkComponent
Human recombinant TFF1
Human recombinant TFF1
Supplier:  Biorbyt
Certificates
Human recombinant TFF1
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    The protein is stable at −20 °C. to −70 °C. in a manual defrost freezer.Avoid repeated freeze thaw cycles
  • Gene ID:
    7031
  • Protein synonyms:
    Trefoil factor 1
  • Protein/peptide name:
    TFF1
  • Purity:
    Greater than 95% by SDS-PAGE gel analysis.
  • Formulation:
    PBS, pH 7.2, sterile filtered
  • Tested applications:
    ELISA , WB , DOT , FuncS

Specifications

Related Information

Frequently Bought Together