Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant DKK1
 :  Biorbyt
 
Human recombinant DKK1
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    The protein is stable at −20 °C. to −70 °C. in a manual defrost freezer.Avoid repeated freeze thaw cycles
  • Protein/peptide name:
    DKK1
  • Purity:
    Greater than 95% by SDS-PAGE gel analysis.
  • Formulation:
    PBS, pH 7.2, sterile filtered
  • Tested applications:
    ELISA , WB , DOT , FuncS

 

 

Frequently Bought Together