Human recombinant GFRalpha 1 (from HEK293 cells)
: Biorbyt
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:GDNF Family Receptor Alpha1
- UniProtKB:P56159 (Human)
- Protein/peptide name:GFRalpha 1
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:DRLDCVKASDQCLKEQSCSTKYRTLRQCVAGKETNFSLASGLEAKDECRSAMEALKQKSLYNCRC KRGMKKEKNCLRIYWSMYQSLQGNDLLEDSPYEPVNSRLSDIFRVVPFISVEHIPKGNNCLDAAK ACNLDDICKKYRSAYITPCTTSVSNDVCNRRKCHKALRQFFDKVPAKHSYGMLFCSCRDIACTER RRQTIVPVCSYEEREKPNCLNLQDSCKTNYICRSRLADFFTNCQPESRSVSSCLKENYADCLLAY SGLIGTVMTPNYIDSSSLSVAPWCDCSNSGNDLEECLKFLNFFKDNTCLKNAIQAFGNGSDVTVW QPAFPVQTTTATTTTALRVKNKPLGPAGSENEIPTHVLPPCANLQAQKLKSNVSGNTHLCISNGN YEKEGLGASSHITTKVDHHHHHH
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2.
- Tested applications:SDS-PAGE
Frequently Bought Together