Human recombinant FH (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Fumarate Hydratase Mitochondrial
- UniProtKB:P07954 (Human)
- Protein/peptide name:FH
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MASQNSFRIEYDTFGELKVPNDKYYGAQTVRSTMNFKIGGVTERMPTPVIKAFGILKRAAAEVNQ DYGLDPKIANAIMKAADEVAEGKLNDHFPLVVWQTGSGTQTNMNVNEVISNRAIEMLGGELGSKI PVHPNDHVNKSQSSNDTFPTAMHIAAAIEVHEVLLPGLQKLHDALDAKSKEFAQIIKIGRTHTQD AVPLTLGQEFSGYVQQVKYAMTRIKAAMPRIYELAAGGTAVGTGLNTRIGFAEKVAAKVAALTGL PFVTAPNKFEALAAHDALVELSGAMNTTACSLMKIANDIRFLGSGPRSGLGELILPENEPGSSIM PGKVNPTQCEAMTMVAAQVMGNHVAVTVGGSNGHFELNVFKPMMIKNVLHSARLLGDASVSFTEN CVVGIQANTERINKLMNESLMLVTALNPHIGYDKAAKIAKTAHKNGSTLKETAIELGYLTAEQFD EWVKPKDMLGPK
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris, pH 8.0.
- Tested applications:SDS-PAGE
Frequently Bought Together