Order Entry
Italy
ContactUsLinkComponent
Human recombinant EPT1 (from E. coli)
Human recombinant EPT1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant EPT1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Ethanolaminephosphotransferase 1
  • UniProtKB:
    Q9C0D9 (Human)
  • Protein/peptide name:
    EPT1
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKL TQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEML KMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKS SKYIAWPLQGWQATFGGGDHPPKSDLVPRGSPEFHMAGYEYVSPEQLAGFDKYKYSAVDTNPLSL YVMHPFWNTIVKVFPTWLAP
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, pH 8.0.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together