Human recombinant IPP1 (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Protein Phosphatase 1 Regulatory Subunit 1A
- UniProtKB:Q13522 (Human)
- Protein/peptide name:IPP1
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MEQDNSPQKIQFTVPLLEPHLDPEAAEQIRRRRPTPATLVLTSDQSSPEIDEDRIPNPHLKSTLA MSPRQRKKMTRITPTMKELQMMVEHHLGQQQQGEEPEGAAESTGTQESRPPGIPDTEVESRLGTS GTAKKTAECIPKTHERGSKEPSTKEPSTHIPPLDSKGANSVLEHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 1mM DTT, 50% Glycerol, pH 8.5.
- Tested applications:SDS-PAGE
Frequently Bought Together