Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant NPPB (from E. coli)
 :  Biorbyt
 
Human recombinant NPPB (from E. coli)
 :  Biorbyt

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Natriuretic Peptides B
  • UniProtKB:
    P16860 (Human)
  • Protein/peptide name:
    NPPB
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMHPLGSPGSASDLETSGLQEQRNHLQGKLSELQVEQTSLEPLQES PRPTGVWKSREVATEGIRGHRKMVLYTLRAPRSPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM EDTA, 1mM DTT, 20% Glycerol, pH 7.4.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together