Recombinant IFN beta 1a
Catalog # BIRBORB97781-10
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Storage conditions:Store at−20 °C.
- Biological activity:EC50: 2.7x108 IU/mg, determined in a viral resistanceassay human "Wish" cell line and VSV virus or themonkey VERO cell line with EMCV virus.
- Protein synonyms:beta-interferon
- UniProtKB:P01574
- Protein/peptide name:IFN beta 1a
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized IFN- insterile bidest water not less than 100 g/ml, which canthen be further diluted to other aqueous solutions.
- Molecular weight:23 kDa
- Formulation:Lyophilized