Order Entry
Italy
ContactUsLinkComponent
Rat IL21 (from E. coli)
Rat IL21 (from E. coli)
Supplier:  Biorbyt
Certificates
Rat IL21 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    Measured by its ability to inhibit IL21dependent proliferation of N1186 human T cells. The ED50 for this effect is typically 1-5ug/mL in the presence of 50 ng/mL of recombinant mouse IL21. Less than 1EU/ug of rRtIL21 as determined by LAL method.
  • Protein synonyms:
    IL 21
  • Protein/peptide name:
    IL21
  • Purity:
    >95% pure by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    15.2 kDa
  • Sequence:
    HKSSPQRPDH LLIRLRHLM D IVEQLKIY EN DLDPELL TAP QDVKGQ CEHE AFACF QKAKL KPSN TGNNKT FIN DLLAQLR RR LPAKRTGN K QRHMAKCPS CDLYEKKTPK EFLERLKWL L QKMIHQHL S
  • Formulation:
    Sterile Filtered White lyophilized (freeze-dried) powder. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together