Human IL8 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:Fully biologically active when compared to standard. Determined by its ability to chemoattract human peripheral blood neutrophils using a concentration range of 10.0-100.0 ng/ml.
- Protein synonyms:310C
- Protein/peptide name:IL8
- Purity:>95% pure by SDS-PAGE and HPLC analyses.
- Molecular weight:8,453 Da
- Sequence:SAKELRCQCI KTYSKPFHPK FIKELRVIES GPHCANTEII VKLSDGRELC LDPKENWVQR VVEKFLKRAE NS
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together