Human IGFBP3 (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50 was determined by its ability to inhibit IGF-II induced proliferation of MCF-7. The expected ED50 for this effect is less than or equal to 0.2 ug/ml in presence of 15 ng/ml of human IGF-II.
- Protein/peptide name:IGFBP3
- Purity:>98% pure by SDS-PAGE and HPLC analyses.
- Molecular weight:28.8 kDa
- Sequence:GASSGGLGPV VRCEPCDARA LAQCAPPPAV CAELVREPGC GCCLTCALSE GQPCGIYTER CGSGLRCQPS PDEARPLQAL LDGRGLCVNA SAVSRLRAYL LPAPPAPGNA SESEEDRSAG EVESPSVSTH RVSDPKFHPL HSKIIIIKKG HAKDSQRYKV DYESQSTDTQ NFSSESKRET EYGPCRREME DTLNHLKFLN VLSPRGVHIP NCDKKGFYKK KQCRPSKGRK RGFCWCVDKY GQPLPGYTTK GKEDHCYS M QSK
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together