Order Entry
Italy
ContactUsLinkComponent
Human IL29 (from E. coli)
Human IL29 (from E. coli)
Supplier:  Biorbyt
Certificates
Human IL29 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    Fully biologically active when compared to standard. The ED50 is determined in an anti-viral assay using human HepG2 cells infected with encephalomyocarditis is typically 1-5 ng/ml.
  • Protein synonyms:
    IFN lambda1
  • Protein/peptide name:
    IL29
  • Purity:
    >97% pure by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    19.8 kDa
  • Sequence:
    PTSKPTTTGK GCHIGRFKSL SPQELASFKK ARDALEESLK LKNWSCSSPV FPGNWDLRLL QVRERPVALE AELALTLKVL EAAAGPALED VLDQPLHTLH HILSQLQACI QPQPTAGPRP RGRLHHWLHR LQEAPKKESA GCLEASVTFN LFRLLTRDLK YVADGNLCLR TSTHPEST
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated (1.0mg/ml) solution in 20mM PB, pH 7.4, 130mM NaCl. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together