Rhesus macaque G-CSF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rhesus monkey
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein/peptide name:G-CSF
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together