Human G-CSF (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:Fully biologically active when compared to standard. The ED50 as calculated by the dose-dependant proliferation of murine NFS-60 indicator cells (measured by 3H-thymidine uptake) is less than 0.1 ng/ml, corresponding to a Specific Activity of 1 x 108 IU/mg. Less than 1EU/ug of rHuG-CSF as determined by LAL method.
- Gene ID:1440
- Protein/peptide name:G-CSF
- Purity:>98% pure by SDS-PAGE and HPLC analyses.
- Molecular weight:18.7 kDa
- Sequence:TPLGPASSLP QSFLLKCLEQ VRKIQGDGAA LQEKLCATYK LCHPEELVLL GHSLGIPWAP LSSCPSQALQ LAGCLSQLHS GLFLYQGLLQ ALEGISPELG PTLDTLQLDV ADFATTIWQQ MEELGMAPAL QPTQGAMPAF ASAFQRRAGG VLVASHLQSF LEVSYRVLRH LAQP
- Formulation:Lyophilized from a 0.2um filtered concentrated (1mg/ml) solution in 10mM sodium acetate buffer pH 4.0. Lyophilized from a 0.2um filtered concentrated (1mg/ml) solution in 10mM sodium acetate buffer pH 4.0.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together