Human recombinant FGF10 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The biological activity was determined by the does-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors yielding an ED50 6 Units/mg. Less than 1EU/ug of rHuKGF-2 as determined by LAL method.
- Protein synonyms:Keratinocye Growth Factor-2
- Protein/peptide name:FGF10
- Purity:>96% pure by SDS-PAGE and HPLC analyses.
- Molecular weight:19.3 kDa
- Sequence:MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
- Formulation:Lyophilized from a 0.2 um filtered concentrated (1mg/ml) solution in PBS, pH 7.4. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together