Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human collagen 4 (from placenta)
Human collagen 4 (from placenta)
BIRBORB81984-0.5
:
Biorbyt
:
Human collagen 4 (from placenta)
BIRBORB81984-0.5
:
Biorbyt
:
Pk:
0,5 mg
Source:
Placenta
Species:
Human
Protein synonyms:
COL4A4
Protein/peptide name:
Collagen 4
Purity:
Purified.
Formulation:
Not lyophilised.
Tested applications:
ELISA