Human Beta-2 gp I (from Plasma)
BIRBORB82010-1
: Biorbyt
:
- Pk:1 mg
- Source:Plasma
- Species:Human
- Protein/peptide name:Beta-2 gp I
- Purity:Purified.
- Formulation:Proprietary formula containing 0.1% preservative, bromo-nitro-dioxane/methylisothiazoline
- Tested applications:ELISA