Human apolipoprotein CII (from Plasma)
Catalog # BIRBORB82147-50
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:50 µG
- Source:Plasma
- Species:Human
- Protein synonyms:Apo CII
- Protein/peptide name:Apolipoprotein CII
- Purity:Purified.
- Formulation:Lyophilised.