Human apolipoprotein B-100 (from Plasma)
BIRBORB82144-0.5
: Biorbyt
:
- Pk:0,5 mg
- Source:Plasma
- Species:Human
- Protein synonyms:Apo B-100
- Protein/peptide name:Apolipoprotein B-100
- Purity:Purified.
- Formulation:Lyophilised.
- Tested applications:SDS-PAGE