Human Recombinant EGF (from P. pastoris)
Catalog # BIRBORB82594-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:P. pastoris
- Species:Human
- Protein synonyms:Epidermal Growth Factor
- Protein/peptide name:EGF
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:WB