West nile virus recombinant west nile virus envelope protein (from E. coli)
Catalog # BIRBORB82559-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:West nile virus
- Protein synonyms:West Nile Virus Envelope
- Protein/peptide name:West Nile Virus Envelope Protein
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB , LF