Recombinant dengue type 2, NS1 protein (from E. coli)
Catalog # BIRBORB82351-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein synonyms:NS1
- Protein/peptide name:Dengue Type 2, NS1 Protein
- Purity:Purified.
- Formulation:Not lyophilised.