Order Entry
Italy
ContactUsLinkComponent
Rubella rubella RSVP (TM) (strain HPV77) >90% viral protein, capsid-free (from vero cells)
Rubella rubella RSVP (TM) (strain HPV77) >90% viral protein, capsid-free (from vero cells)
Catalog # BIRBORB81838-1
Supplier:  Biorbyt
CAS Number:  
Rubella rubella RSVP (TM) (strain HPV77) >90% viral protein, capsid-free (from vero cells)
Catalog # BIRBORB81838-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Vero cells
  • Species:
    Rubella
  • Protein synonyms:
    Rubella RSVP (TM) (Strain HPV77) >90% Viral
  • Protein/peptide name:
    Rubella RSVP (TM) (Strain HPV77) >90% Viral Protein, Capsid-Free
  • Purity:
    >90%
  • Formulation:
    Not lyophilised.
  • Tested applications:
    ELISA , WB
  • Purification:
    Purified

Specifications

About this item